Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: VDR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005636918.2 | XP_005636975.1 | 6 - 7 | 858 | 100 | |
XM_005636919.3 | XP_005636976.1 | 8 - 9 | 998 | 100 | ||
XM_005636920.2 | XP_005636977.1 | 7 - 8 | 938 | 100 |
Set Membership: |
Amplicon length between 86 and 100 Canis familiaris Probe spans exons |