Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: VDR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005636918.2 | XP_005636975.1 | 7 - 8 | 1014 | 83 | |
XM_005636919.3 | XP_005636976.1 | 9 - 10 | 1154 | 83 | ||
XM_005636920.2 | XP_005636977.1 | 8 - 9 | 1094 | 83 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Canis familiaris Probe spans exons |