Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DCT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001284466.1 | NP_001271395.1 | 1 - 2 | 389 | 84 | |
GenBank mRNA | AB766380.1 | - | 1 - 2 | 389 | 84 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Recommended Assay Canis familiaris Probe spans exons |