Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AP3M1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001287115.1 | NP_001274044.1 | 8 - 9 | 1196 | 66 | |
XM_005619059.3 | XP_005619116.1 | 8 - 9 | 1526 | 66 | ||
XM_022417340.1 | XP_022273048.1 | 8 - 9 | 1471 | 66 | ||
GenBank mRNA | DN871041.1 | - | 5 - 6 | 620 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Canis familiaris Probe spans exons |