Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MMP7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001242726.1 | NP_001229655.1 | 4 - 5 | 610 | 58 | |
GenBank mRNA | DN873520.1 | - | 2 - 3 | 458 | 58 | |
DN873521.1 | - | 3 - 4 | 508 | 58 | ||
JF412032.1 | - | 3 - 4 | 513 | 58 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Canis familiaris Inventoried Probe spans exons |