Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CA5B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003640227.3 | XP_003640275.1 | 1 - 2 | 132 | 63 |
Set Membership: |
Amplicon length less than or equal to 70 Canis familiaris Probe spans exons |