Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CA5B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003640227.3 | XP_003640275.1 | 2 - 3 | 333 | 112 | |
XM_022416368.1 | XP_022272076.1 | 6 - 7 | 1261 | 112 | ||
XM_022416369.1 | XP_022272077.1 | 6 - 7 | 912 | 112 | ||
XM_022416370.1 | XP_022272078.1 | 3 - 4 | 398 | 112 | ||
XM_022416371.1 | XP_022272079.1 | 3 - 4 | 305 | 112 |
Set Membership: |
Amplicon length greater than or equal to 101 Canis familiaris Probe spans exons |