Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RWDD2B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_022413312.1 | XP_022269020.1 | 5 - 6 | 997 | 91 | |
XM_022413313.1 | XP_022269021.1 | 5 - 6 | 904 | 91 | ||
XM_022413314.1 | XP_022269022.1 | 5 - 6 | 816 | 91 | ||
XM_022413315.1 | XP_022269023.1 | 5 - 6 | 1092 | 91 | ||
XM_022413316.1 | XP_022269024.1 | 3 - 4 | 471 | 91 | ||
XM_022413317.1 | XP_022269025.1 | 3 - 4 | 410 | 91 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Canis familiaris Probe spans exons |