Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC16A13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_844349.5 | XP_849442.1 | 1 - 2 | 623 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 Canis familiaris Probe spans exons |