Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ECI1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005622087.3 | XP_005622144.1 | 6 - 7 | 896 | 55 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Canis familiaris Probe spans exons |