Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CYBA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001100290.1 | NP_001093760.1 | 129 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 Canis familiaris |