Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GGT6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003434611.4 | XP_003434659.2 | 2 - 3 | 260 | 68 | |
XM_022418401.1 | XP_022274109.1 | 1 - 2 | 660 | 68 | ||
XM_022418402.1 | XP_022274110.1 | 2 - 3 | 263 | 68 | ||
XM_022418403.1 | XP_022274111.1 | 1 - 2 | 662 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Canis familiaris Probe spans exons |