Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZG16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_022419866.1 | XP_022275574.1 | 2 - 3 | 283 | 63 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Canis familiaris Probe spans exons |