Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PYY | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_022422937.1 | XP_022278645.1 | 1275 | 92 |
Set Membership: |
Amplicon length between 86 and 100 Canis familiaris |