Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GAL3ST4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_022419663.1 | XP_022275371.1 | 792 | 95 | ||
XM_022419664.1 | XP_022275372.1 | 825 | 95 | |||
XM_546961.6 | XP_546961.2 | 769 | 95 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Recommended Assay Canis familiaris |