Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SFRP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003639564.2 | XP_003639612.1 | 473 | 69 | ||
GenBank mRNA | AB672664.1 | - | 417 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 G-protein coupled receptor Canis familiaris |