Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC39A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003639152.4 | XP_003639200.1 | 2 - 3 | 334 | 93 | |
XM_005622808.3 | XP_005622865.1 | 2 - 3 | 314 | 93 | ||
XM_022421392.1 | XP_022277100.1 | 2 - 3 | 374 | 93 |
Set Membership: |
Amplicon length between 86 and 100 Canis familiaris Probe spans exons |