Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TXNDC17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005620315.3 | XP_005620372.1 | 2 - 3 | 323 | 72 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Recommended Assay Canis familiaris Probe spans exons |