Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MYL6B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_014117152.2 | XP_013972627.1 | 581 | 81 | ||
XM_843490.5 | XP_848583.1 | 548 | 81 |
Set Membership: |
Amplicon length between 71 and 85 Canis familiaris |