Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IDS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001048125.1 | NP_001041590.1 | 2 - 3 | 238 | 73 | |
XM_005641023.3 | XP_005641080.1 | 2 - 3 | 385 | 73 |
Set Membership: |
Amplicon length between 71 and 85 Canis familiaris Probe spans exons |