Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ANKH | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005619428.3 | XP_005619485.1 | 3 - 4 | 742 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 Canis familiaris Probe spans exons |