Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RASA4B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005620995.3 | XP_005621052.1 | 346 | 68 | ||
XM_014114180.2 | XP_013969655.1 | 346 | 68 | |||
XM_022419597.1 | XP_022275305.1 | 346 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 Canis familiaris |