Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DDIT3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005625542.3 | XP_005625599.2 | 319 | 145 | ||
XM_014117187.2 | XP_013972662.1 | 364 | 145 | |||
XM_022424097.1 | XP_022279805.1 | 390 | 145 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Recommended Assay Canis familiaris Inventoried |