Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BNIP3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_022412057.1 | XP_022267765.1 | 2 - 3 | 219 | 87 | |
XM_844054.6 | XP_849147.4 | 2 - 3 | 330 | 87 | ||
GenBank mRNA | DQ926865.1 | - | 2 - 3 | 195 | 87 |
Set Membership: |
Amplicon length between 86 and 100 Canis familiaris Probe spans exons |