Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SURF2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_548389.6 | XP_548389.2 | 2 - 3 | 310 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Canis familiaris Probe spans exons |