Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SURF2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_548389.6 | XP_548389.2 | 3 - 4 | 406 | 63 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Canis familiaris Probe spans exons |