Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DPT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001287158.1 | NP_001274087.1 | 1 - 2 | 348 | 65 | |
GenBank mRNA | DN379133.1 | - | 1 - 2 | 348 | 65 | |
HM775211.1 | - | 1 - 2 | 252 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 Canis familiaris Probe spans exons |