Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NPTX2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003639083.4 | XP_003639131.1 | 1086 | 83 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Canis familiaris |