Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RAB25 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001313865.1 | NP_001300794.1 | 3 - 4 | 698 | 58 | |
XM_014114978.2 | XP_013970453.1 | 3 - 4 | 765 | 58 | ||
XM_022420762.1 | XP_022276470.1 | 3 - 4 | 770 | 58 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Canis familiaris Probe spans exons |