Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CRYBA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001080899.1 | NP_001074368.1 | 2 - 3 | 93 | 85 | |
GenBank mRNA | EF191198.1 | - | 2 - 3 | 93 | 85 |
Set Membership: |
Amplicon length between 71 and 85 Canis familiaris Probe spans exons |