Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: APEX2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003640275.4 | XP_003640323.1 | 5 - 6 | 1171 | 75 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Canis familiaris Probe spans exons |