Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LNP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005639396.3 | XP_005639453.1 | 386 | 68 | ||
XM_022414016.1 | XP_022269724.1 | 299 | 68 | |||
XM_022414017.1 | XP_022269725.1 | 318 | 68 | |||
XM_022414018.1 | XP_022269726.1 | 284 | 68 | |||
XM_549598.7 | XP_549598.2 | 411 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Canis familiaris |