Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: B4GALT4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001127498.1 | NP_001120970.1 | 682 | 73 | ||
GenBank mRNA | AM989461.1 | - | 682 | 73 |
Set Membership: |
Amplicon length between 71 and 85 Canis familiaris |