Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: B4GALT4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001127498.1 | NP_001120970.1 | 870 | 126 | ||
GenBank mRNA | AM989461.1 | - | 870 | 126 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Canis familiaris |