Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MED21 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001197311.1 | NP_001184240.1 | 2 - 3 | 179 | 124 | |
XM_005636700.3 | XP_005636757.1 | 2 - 3 | 252 | 124 | ||
GenBank mRNA | GU811151.1 | - | 2 - 3 | 179 | 124 |
Set Membership: |
Amplicon length greater than or equal to 101 Canis familiaris Probe spans exons |