Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PXMP4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005634946.3 | XP_005635003.1 | 1 - 2 | 195 | 96 | |
XM_847386.5 | XP_852479.2 | 1 - 2 | 190 | 96 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Canis familiaris Probe spans exons |