Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RBBP9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_844630.5 | XP_849723.3 | 2 - 3 | 217 | 101 |
Set Membership: |
Amplicon length greater than or equal to 101 Canis familiaris Probe spans exons |