Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MED28 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005618542.3 | XP_005618599.1 | 3 - 4 | 365 | 84 | |
XM_022417147.1 | XP_022272855.1 | 3 - 4 | 365 | 84 | ||
XM_845684.4 | XP_850777.1 | 3 - 4 | 365 | 84 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Canis familiaris Probe spans exons |