Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RAB31 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_022421633.1 | XP_022277341.1 | 4 - 5 | 401 | 90 | |
XM_845517.5 | XP_850610.2 | 5 - 6 | 489 | 90 |
Set Membership: |
Amplicon length between 86 and 100 Canis familiaris Probe spans exons |