Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HYLS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005619586.2 | XP_005619643.1 | 130 | 75 | ||
XM_022418112.1 | XP_022273820.1 | 277 | 75 | |||
XM_846949.5 | XP_852042.1 | 155 | 75 |
Set Membership: |
Amplicon length between 71 and 85 Canis familiaris |