Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: POLR3G | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005618122.3 | XP_005618179.1 | 426 | 122 | ||
XM_005618123.3 | XP_005618180.1 | 426 | 122 | |||
XM_014112225.2 | XP_013967700.1 | 298 | 122 | |||
XM_014112227.2 | XP_013967702.1 | 298 | 122 | |||
XM_847057.5 | XP_852150.1 | 386 | 122 |
Set Membership: |
Amplicon length greater than or equal to 101 Canis familiaris |