Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SMTNL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_022405409.1 | XP_022261117.1 | 4 - 5 | 1191 | 88 | |
XM_022405410.1 | XP_022261118.1 | 3 - 4 | 1056 | 88 | ||
XM_540610.6 | XP_540610.2 | 3 - 4 | 1020 | 88 |
Set Membership: |
Amplicon length between 86 and 100 Canis familiaris Probe spans exons |