Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RCAN2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_014118488.2 | XP_013973963.1 | 2 - 3 | 401 | 85 | |
XM_014118489.2 | XP_013973964.1 | 2 - 3 | 458 | 85 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Recommended Assay Canis familiaris Probe spans exons |