Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PRAP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005637911.3 | XP_005637968.1 | 2 - 3 | 210 | 142 | |
XM_845222.5 | XP_850315.1 | 2 - 3 | 210 | 142 |
Set Membership: |
Amplicon length greater than or equal to 101 Canis familiaris Probe spans exons |