Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLX1A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005621358.3 | XP_005621415.1 | 3 - 4 | 599 | 73 | |
XM_844501.5 | XP_849594.1 | 3 - 4 | 599 | 73 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Canis familiaris Probe spans exons |