Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: WRAP53 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005619990.3 | XP_005620047.1 | 7 - 8 | 2326 | 91 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Canis familiaris Probe spans exons |