Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GPR89A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_022404931.1 | XP_022260639.1 | 8 - 9 | 893 | 107 | |
XM_844483.5 | XP_849576.1 | 8 - 9 | 893 | 107 |
Set Membership: |
Amplicon length greater than or equal to 101 G-protein coupled receptor Canis familiaris Probe spans exons |