Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GPR89A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005630713.3 | XP_005630770.1 | 10 - 11 | 1097 | 157 | |
XM_022404932.1 | XP_022260640.1 | 9 - 10 | 993 | 157 | ||
XM_844483.5 | XP_849576.1 | 10 - 11 | 1079 | 157 |
Set Membership: |
Amplicon length greater than or equal to 101 G-protein coupled receptor Canis familiaris Probe spans exons |