Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CRYBB2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001048113.1 | NP_001041578.1 | 2 - 3 | 102 | 100 | |
GenBank mRNA | DQ324463.1 | - | 2 - 3 | 102 | 100 |
Set Membership: |
Amplicon length between 86 and 100 Canis familiaris Probe spans exons |