Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: B2M | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003640047.2 | XP_003640095.1 | 3 - 4 | 588 | 92 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Canis familiaris Inventoried Probe spans exons |